Learn More
Syngene™ AirClean™ Mecury Vapor Filter
GREENER_CHOICE
Product Code. 17705964 Shop All Syngene Products
Change view
Click to view available options
Width (Metric):
46 cm
61 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
17705964 46 cm Each
17795954 61 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17705964 Supplier Syngene™ Supplier No. SYNACF43018
The AirClean™ Mecury Vapor Filter is used to absorb fumes and odors.
  • These filters are impregnated with a chemical additive which helps the adsorption process by chemically or physically reacting with the toxic contaminant.
Specifications
Length (English) 12 in.
Depth (English) 3 in.
Width (English) 18 in.
Depth (Metric) 8 cm
Length (Metric) 30.5 cm
Width (Metric) 46 cm
For Use With (Application) Mercury Vapor