Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UF55, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ UF55B02072
Product Code. 17589445
  • ONLINE EXCLUSIVE PRICE
    87.20€
    84.00€ / Each
    Save 3.20€
    3% Off
Only null left
Estimated Shipment: 16-12-2025
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
UF55 Oven
Drip Tray
Stainless Steel
Rim: 15 mm