Learn More
BUCHI Pfeiffer Duo 6 Vacuum Pump for Lyovapor Freeze Dryer
The DuoVane series consists of innovative two-stage oil-lubricated rotary vane pumps that are ideal for use with Lyovapor Freeze Dryers
Brand:  BUCHI 11065652
Product Code. 17294654
  • ONLINE EXCLUSIVE PRICE
    4070.00€
    3605.52€ / Each
    Save 464.48€
    11% Off
Only null left
Estimated Shipment: 24-11-2025
Description
  • 6 m3/h 60 Hz pump speed range
  • For Lyovapor L-200/L-250
Specifications
Specifications
1500 hPa
5 m³/hr.
Rotary Vane Vacuum Pump
230/240 V