Learn More
Labconco High Capacity Micro and 12 x 32 Rotor - CentriVap Pro
Shop All Labconco Corporation Products
Description
- Compatible with CentriVap Pro Vacuum Concentrators
- Stackable design
- Stainless steel material\
- Optional PTFE coated
Specifications
Specifications
| Material | Stainless Steel |
| Type | Rotor |
| Material (Coating) | PTFE |
| Rotor System | CentriVap Pro |
| Height (English) | 1.8 in. |
| Height (Metric) | 4.57 cm |
| Diameter (English) | 11.4 in. |
| Diameter (Metric) | 28.96 cm |