Learn More
Memmert™ Entry Port
Product Code. 15652234 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
ICO50
Unit Size:
Each
Material: Silicone; For Use With: ICO50
  • Moisture tight
  • Can be closed by silicone stopper
Specifications
For Use With (Equipment) ICO50
Material Silicone
Type Entry Port
Diameter (Metric) Inside 40 mm