Learn More
Mölnlycke™ Biogel™ PI Micro Tech Indicator Underglove
Sterile, blue cleanroom undergloves with beaded cuffs and curved fingers are made to be worn in combination with the Biogel™ PI Micro Tech overglove to form a breach indication system. The gloves are suitable for use in aseptic and Class 100 (ISO 5) / EU GMP Grade A environments.
296.64€ - 370.24€
Specifications
| Certifications/Compliance | ASTM D3577 Viral penetration ASTM F1671 Pyrogenicity ASTM D7102 |
|---|---|
| Certifications/Compliance (Europe) | EN 455, ISO 10282 , EN 374, EN 16523-1, PPE Cat. III, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, Allergenicity ISO 10993, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420 |
| Cleanroom Class | Class 100 (ISO 5) |
| Accelerator Free | No |
| AQL | 0.65 |
| Product Code | Brand | Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Size | Price | Quantity & Availability | |||||
17885748
![]() |
Mölnlycke™
4448955-00 |
5.5 |
556.50€ 370.24€ / 200 pairs Save 186.26€ 33% Off |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||||
17865758
![]() |
Mölnlycke™
4448960-00 |
6 |
556.50€ 370.24€ / 200 pairs Save 186.26€ 33% Off |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||||
17855778
![]() |
Mölnlycke™
4448965-00 |
6.5 |
556.50€ 370.24€ / 200 pairs Save 186.26€ 33% Off |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||||
17895758
![]() |
Mölnlycke™
4448970-00 |
7 |
556.50€ 370.24€ / 200 pairs Save 186.26€ 33% Off |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||||
17875748
![]() |
Mölnlycke™
4448975-00 |
7.5 |
556.50€ 370.24€ / 200 pairs Save 186.26€ 33% Off |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||||
17815758
![]() |
Mölnlycke™
4448980-00 |
8 |
556.50€ 370.24€ / 200 pairs Save 186.26€ 33% Off |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||||
17825758
![]() |
Mölnlycke™
4448985-00 |
8.5 |
556.50€ 370.24€ / 200 pairs Save 186.26€ 33% Off |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||||
17875778
![]() |
Mölnlycke™
4448990-00 |
9 |
445.20€ 296.64€ / 160 pairs Save 148.56€ 33% Off |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||||
Description
- Blue underglove can be worn together with Biogel™ PI Micro Tech to form a breach indication system
- Tested and cleared for use with chemotherapy agents
- Synthetic polyisoprene
- Exceed industry standard for force at break, tensile strength and elongation
- Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
Specifications
| ASTM D3577 Viral penetration ASTM F1671 Pyrogenicity ASTM D7102 | |
| Class 100 (ISO 5) | |
| 0.65 | |
| Single-use | |
| 8.3 mil | |
| Specific | |
| Polyisoprene | |
| 0.19 mm | |
| Sterile | |
| 0.16 mm | |
| 11.8 in. |
| EN 455, ISO 10282 , EN 374, EN 16523-1, PPE Cat. III, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, Allergenicity ISO 10993, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420 | |
| No | |
| Blue | |
| Smooth | |
| 0.21 mm | |
| Yes | |
| 7.5 mil | |
| Powder Free | |
| 6.3 mil (Cuff) | |
| 24 MPa | |
| 30 cm |