Learn More
BUCHI Beaker and Drying Flasks for Rotary Evaporators
Special shaped flasks for large samples and powder drying.
677.00 DKK - 902.00 DKK
| Product Code | Brand | Type | Volume (Metric) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Type | Volume (Metric) | Price | Quantity & Availability | ||||
|
15510590
|
BUCHI
11063158 |
Drying Flask | 500 mL |
677.00 DKK
Each |
Estimated Shipment: 05-12-2025
Log in to see stock. |
||||
|
15520590
|
BUCHI
11063160 |
Drying Flask | 1500 mL |
757.00 DKK
Each |
Estimated Shipment: 17-11-2025
Log in to see stock. |
||||
|
15590580
|
BUCHI
11063154 |
Beaker Flask | 500 mL |
737.00 DKK
Each |
Estimated Shipment: 05-12-2025
Log in to see stock. |
||||
|
17214664
|
BUCHI
11065690 |
Beaker Flask | 4000 mL |
902.00 DKK
Each |
Estimated Shipment: 05-12-2025
Log in to see stock. |
||||
|
17224664
|
BUCHI
11065718 |
Beaker Flask | 1500 mL |
882.00 DKK
Each |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||||
Beskrivning
- For use with the rotary evaporators
- Available in multiple sizes with different standard joints
Specifikationer
| Borosilicate Glass (3.3) |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel